Assay Details
| Gene Symbol: PRAF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_106346.3 | NP_565144.1 | 5 - 6 | 946 | 58 | |
| GenBank mRNA | AF323270.1 | - | 5 - 6 | 900 | 58 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

PRAF1| Gene Symbol: PRAF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_106346.3 | NP_565144.1 | 5 - 6 | 946 | 58 | |
| GenBank mRNA | AF323270.1 | - | 5 - 6 | 900 | 58 | |
Set Membership: |
|