Assay Details
| Gene Symbol: AT1G77100 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_106360.3 | NP_177835.3 | 1 - 2 | 208 | 71 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AT1G77100| Gene Symbol: AT1G77100 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_106360.3 | NP_177835.3 | 1 - 2 | 208 | 71 | |
Set Membership: |
|