Assay Details
| Gene Symbol: AT1G77310 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001334775.1 | NP_001321460.1 | 1 - 2 | 419 | 139 | |
| NM_106380.7 | NP_177855.4 | 1 - 2 | 426 | 139 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001334775.1 transcripts.
Set Membership: |
|

AT1G77310
Genomic Map