Assay Details
| Gene Symbol: CYCA1;2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_106388.4 | NP_177863.2 | 7 - 8 | 1132 | 64 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

CYCA1;2| Gene Symbol: CYCA1;2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_106388.4 | NP_177863.2 | 7 - 8 | 1132 | 64 | |
Set Membership: |
|