Assay Details
| Gene Symbol: BGAL16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001334779.1 | NP_001320598.1 | 16 - 17 | 2186 | 127 | |
| NM_001334780.1 | NP_001320597.1 | 17 - 18 | 2108 | 127 | ||
| NM_001334781.1 | NP_001320596.1 | 14 - 15 | 1682 | 127 | ||
| NM_106391.2 | NP_177866.2 | 17 - 18 | 2093 | 127 | ||
More Information
Set Membership: |
|

BGAL16
Genomic Map