Assay Details
| Gene Symbol: BGAL16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001334779.1 | NP_001320598.1 | 1 - 2 | 271 | 80 | |
| NM_001334780.1 | NP_001320597.1 | 1 - 2 | 271 | 80 | ||
| NM_106391.2 | NP_177866.2 | 1 - 2 | 271 | 80 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001334779.1 transcripts.
Set Membership: |
|

BGAL16
Genomic Map