Assay Details
| Gene Symbol: BGAL16 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001334780.1 | NP_001320597.1 | 3 - 4 | 489 | 84 | |
| NM_106391.2 | NP_177866.2 | 3 - 4 | 489 | 84 | ||
More Information
Set Membership: |
|

BGAL16
Genomic Map