Assay Details
| Gene Symbol: RFC3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_106396.5 | NP_177871.1 | 7 - 8 | 1158 | 71 | |
| GenBank mRNA | AY065452.1 | - | 7 - 8 | 1008 | 71 | |
| AY117282.1 | - | 7 - 8 | 933 | 71 | ||
More Information
Set Membership: |
|

RFC3
Genomic Map