Assay Details
| Gene Symbol: TAPX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001334784.1 | NP_001321064.1 | 2 - 3 | 463 | 95 | |
| NM_106398.3 | NP_177873.1 | 2 - 3 | 505 | 95 | ||
| GenBank mRNA | AK229693.1 | - | 2 - 3 | 381 | 95 | |
| AY085554.1 | - | 2 - 3 | 395 | 95 | ||
More Information
Set Membership: |
|

TAPX
Genomic Map