Assay Details
| Gene Symbol: AT1G77550 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_106404.6 | NP_177879.3 | 7 - 8 | 1415 | 100 | |
| GenBank mRNA | AK226662.1 | - | 7 - 8 | 1382 | 100 | |
| BT014678.1 | - | 7 - 8 | 1330 | 100 | ||
More Information
Set Membership: |
|

AT1G77550
Genomic Map