Assay Details
| Gene Symbol: AT1G77620 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_106410.3 | NP_177885.1 | 12 - 13 | 1931 | 92 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AT1G77620| Gene Symbol: AT1G77620 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_106410.3 | NP_177885.1 | 12 - 13 | 1931 | 92 | |
Set Membership: |
|