Assay Details
| Gene Symbol: AGL67 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001124144.1 | NP_001117616.1 | 1 - 2 | 182 | 68 | |
| NM_001334806.1 | NP_001322357.1 | 2 - 3 | 226 | 68 | ||
| NM_001334807.1 | NP_001322358.1 | 1 - 2 | 306 | 68 | ||
| NM_106444.3 | NP_177918.2 | 1 - 2 | 306 | 68 | ||
More Information
Set Membership: |
|

AGL67
Genomic Map