Assay Details
| Gene Symbol: AGL67 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001124144.1 | NP_001117616.1 | 3 - 4 | 269 | 102 | |
| NM_001334806.1 | NP_001322357.1 | 4 - 5 | 313 | 102 | ||
| NM_001334807.1 | NP_001322358.1 | 3 - 4 | 393 | 102 | ||
| NM_106444.3 | NP_177918.2 | 3 - 4 | 393 | 102 | ||
More Information
Set Membership: |
|

AGL67
Genomic Map