Assay Details
| Gene Symbol: AST56 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_106448.4 | NP_565165.2 | 8 - 9 | 1618 | 86 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

AST56| Gene Symbol: AST56 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_106448.4 | NP_565165.2 | 8 - 9 | 1618 | 86 | |
Set Membership: |
|