Assay Details
| Gene Symbol: UGT85A4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_106476.3 | NP_177950.1 | 1 - 2 | 712 | 144 | |
| GenBank mRNA | AY099642.1 | - | 1 - 2 | 651 | 144 | |
| BT000242.1 | - | 1 - 2 | 518 | 144 | ||
| KJ138927.1 | - | 1 - 2 | 518 | 144 | ||
More Information
Set Membership: |
|

UGT85A4
Genomic Map