Assay Details
| Gene Symbol: SNRK2-8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001084370.2 | NP_001077839.1 | 6 - 7 | 977 | 99 | |
| NM_202441.3 | NP_974170.1 | 5 - 6 | 1324 | 99 | ||
| GenBank mRNA | AF411782.1 | - | 5 - 6 | 956 | 99 | |
| BT020608.1 | - | 5 - 6 | 740 | 99 | ||
More Information
Set Membership: |
|

SNRK2-8
Genomic Map