Assay Details
| Gene Symbol: SHB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198498.1 | NP_001185427.1 | 3 - 4 | 770 | 78 | |
| NM_001334830.1 | NP_001322353.1 | 3 - 4 | 868 | 78 | ||
| NM_106501.5 | NP_177975.3 | 3 - 4 | 868 | 78 | ||
| GenBank mRNA | AK221532.1 | - | 3 - 4 | 758 | 78 | |
More Information
Set Membership: |
|

SHB
Genomic Map