Assay Details
| Gene Symbol: NADK3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001334835.1 | NP_001321203.1 | 2 - 3 | 428 | 101 | |
| NM_106506.3 | NP_177980.2 | 2 - 3 | 436 | 101 | ||
| GenBank mRNA | BT022078.1 | - | 2 - 3 | 358 | 101 | |
| BT033136.1 | - | 2 - 3 | 334 | 101 | ||
More Information
Set Membership: |
|

NADK3
Genomic Map