Assay Details
| Gene Symbol: NADK3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001334835.1 | NP_001321203.1 | 3 - 4 | 513 | 64 | |
| NM_106506.3 | NP_177980.2 | 3 - 4 | 521 | 64 | ||
| GenBank mRNA | BT022078.1 | - | 3 - 4 | 443 | 64 | |
| BT033136.1 | - | 3 - 4 | 419 | 64 | ||
More Information
Set Membership: |
|

NADK3
Genomic Map