Assay Details
| Gene Symbol: GGH3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_106514.4 | NP_177988.1 | 8 - 9 | 1077 | 83 | |
| GenBank mRNA | AF389281.1 | - | 8 - 9 | 967 | 83 | |
| BT000554.1 | - | 8 - 9 | 928 | 83 | ||
More Information
Set Membership: |
|

GGH3
Genomic Map