Assay Details
| Gene Symbol: GGH3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_106514.4 | NP_177988.1 | 9 - 10 | 1179 | 88 | |
| GenBank mRNA | AF389281.1 | - | 9 - 10 | 1069 | 88 | |
More Information
Set Membership: |
|

GGH3
Genomic Map