Assay Details
| Gene Symbol: APC6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_106524.2 | NP_565188.1 | 12 - 13 | 1782 | 99 | |
| GenBank mRNA | BT033023.1 | - | 12 - 13 | 1280 | 99 | |
More Information
Set Membership: |
|

APC6
Genomic Map