Assay Details
| Gene Symbol: APC6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_106524.2 | NP_565188.1 | 1 - 2 | 745 | 86 | |
| GenBank mRNA | BT033023.1 | - | 1 - 2 | 243 | 86 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

APC6| Gene Symbol: APC6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_106524.2 | NP_565188.1 | 1 - 2 | 745 | 86 | |
| GenBank mRNA | BT033023.1 | - | 1 - 2 | 243 | 86 | |
Set Membership: |
|