Assay Details
| Gene Symbol: AT1G78800 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_106527.4 | NP_178001.2 | 7 - 8 | 998 | 77 | |
| GenBank mRNA | KJ138925.1 | - | 7 - 8 | 915 | 77 | |
More Information
Set Membership: |
|

AT1G78800
Genomic Map