Assay Details
| Gene Symbol: VP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001124147.1 | NP_001117619.1 | 13 - 14 | 2545 | 77 | |
| NM_001334854.1 | NP_001322698.1 | 13 - 14 | 2560 | 77 | ||
| NM_106541.4 | NP_565195.1 | 13 - 14 | 2627 | 77 | ||
| GenBank mRNA | AB034696.1 | - | 13 - 14 | 2465 | 77 | |
| AY054485.1 | - | 13 - 14 | 2545 | 77 | ||
| BT010353.1 | - | 12 - 13 | 2094 | 77 | ||
More Information
Set Membership: |
|

VP2
Genomic Map