Assay Details
| Gene Symbol: VP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001124147.1 | NP_001117619.1 | 5 - 6 | 1060 | 66 | |
| NM_001334854.1 | NP_001322698.1 | 5 - 6 | 1075 | 66 | ||
| NM_106541.4 | NP_565195.1 | 5 - 6 | 1142 | 66 | ||
| GenBank mRNA | AB034696.1 | - | 5 - 6 | 980 | 66 | |
| AY054485.1 | - | 5 - 6 | 1060 | 66 | ||
| BT010353.1 | - | 4 - 5 | 609 | 66 | ||
More Information
Set Membership: |
|

VP2
Genomic Map