Assay Details
| Gene Symbol: SRF5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_106547.3 | NP_178019.2 | 12 - 13 | 1713 | 142 | |
| GenBank mRNA | AY518290.1 | - | 12 - 13 | 1713 | 142 | |
| FJ708683.1 | - | 12 - 13 | 1558 | 142 | ||
More Information
Set Membership: |
|

SRF5
Genomic Map