Assay Details
| Gene Symbol: EMB1135 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_106583.6 | NP_178053.4 | 5 - 6 | 1427 | 99 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

EMB1135| Gene Symbol: EMB1135 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_106583.6 | NP_178053.4 | 5 - 6 | 1427 | 99 | |
Set Membership: |
|