Assay Details
| Gene Symbol: SMB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001334893.1 | NP_001319416.1 | 3 - 4 | 756 | 118 | |
| NM_001334894.1 | NP_001323331.1 | 2 - 3 | 1036 | 118 | ||
| NM_106606.2 | NP_178076.1 | 3 - 4 | 757 | 118 | ||
| NM_202449.2 | NP_974178.1 | 3 - 4 | 767 | 118 | ||
| NM_202450.2 | NP_974179.1 | 3 - 4 | 708 | 118 | ||
More Information
Set Membership: |
|

SMB
Genomic Map