Assay Details
| Gene Symbol: GC5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001124150.1 | NP_001117622.1 | 3 - 4 | 1247 | 69 | |
| NM_001198512.1 | NP_001185441.1 | 3 - 4 | 1360 | 69 | ||
| NM_001198513.2 | NP_001185442.1 | 3 - 4 | 1360 | 69 | ||
| NM_001334925.1 | NP_001321438.1 | 3 - 4 | 1366 | 69 | ||
| NM_106632.4 | NP_178101.3 | 3 - 4 | 1347 | 69 | ||
| GenBank mRNA | AK226727.1 | - | 3 - 4 | 1249 | 69 | |
| EU249329.1 | - | 2 - 3 | 1090 | 69 | ||
More Information
Set Membership: |
|

GC5
Genomic Map