Assay Details
| Gene Symbol: GC5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001124150.1 | NP_001117622.1 | 7 - 8 | 1577 | 83 | |
| NM_001198512.1 | NP_001185441.1 | 7 - 8 | 1690 | 83 | ||
| NM_001198513.2 | NP_001185442.1 | 7 - 8 | 1690 | 83 | ||
| NM_001334925.1 | NP_001321438.1 | 7 - 8 | 1696 | 83 | ||
| NM_106632.4 | NP_178101.3 | 7 - 8 | 1677 | 83 | ||
| GenBank mRNA | AK226727.1 | - | 7 - 8 | 1579 | 83 | |
| EU249329.1 | - | 6 - 7 | 1420 | 83 | ||
More Information
Set Membership: |
|

GC5
Genomic Map