Assay Details
| Gene Symbol: GC5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001124150.1 | NP_001117622.1 | 9 - 10 | 1902 | 80 | |
| NM_001198512.1 | NP_001185441.1 | 9 - 10 | 2015 | 80 | ||
| NM_001198513.2 | NP_001185442.1 | 9 - 10 | 2015 | 80 | ||
| NM_001334925.1 | NP_001321438.1 | 9 - 10 | 2021 | 80 | ||
| NM_106632.4 | NP_178101.3 | 9 - 10 | 2002 | 80 | ||
| GenBank mRNA | AK226727.1 | - | 9 - 10 | 1904 | 80 | |
| EU249329.1 | - | 8 - 9 | 1745 | 80 | ||
More Information
Set Membership: |
|

GC5
Genomic Map