Assay Details
| Gene Symbol: GL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198514.1 | NP_001185443.1 | 8 - 9 | 2004 | 84 | |
| NM_001334926.1 | NP_001320267.1 | 7 - 8 | 2106 | 84 | ||
| NM_001334927.1 | NP_001320268.1 | 7 - 8 | 1875 | 84 | ||
| NM_001334929.1 | NP_001320270.1 | 6 - 7 | 1768 | 84 | ||
| NM_106633.2 | NP_565223.1 | 7 - 8 | 1930 | 84 | ||
| GenBank mRNA | AF360294.1 | - | 7 - 8 | 1891 | 84 | |
| AJ457046.1 | - | 7 - 8 | 1780 | 84 | ||
| BT001956.1 | - | 7 - 8 | 1780 | 84 | ||
More Information
Set Membership: |
|

GL2
Genomic Map