Assay Details
| Gene Symbol: GL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198514.1 | NP_001185443.1 | 9 - 10 | 2085 | 65 | |
| NM_001334926.1 | NP_001320267.1 | 8 - 9 | 2187 | 65 | ||
| NM_001334929.1 | NP_001320270.1 | 7 - 8 | 1849 | 65 | ||
| NM_106633.2 | NP_565223.1 | 8 - 9 | 2011 | 65 | ||
| GenBank mRNA | AF360294.1 | - | 8 - 9 | 1972 | 65 | |
| AJ457046.1 | - | 8 - 9 | 1861 | 65 | ||
| BT001956.1 | - | 8 - 9 | 1861 | 65 | ||
More Information
Set Membership: |
|

GL2
Genomic Map