Assay Details
| Gene Symbol: La2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001334930.1 | NP_001319421.1 | 3 - 4 | 565 | 82 | |
| NM_106637.5 | NP_178106.2 | 4 - 5 | 490 | 82 | ||
| NM_202456.2 | NP_974185.1 | 3 - 4 | 524 | 82 | ||
| GenBank mRNA | AK319137.1 | - | 3 - 4 | 525 | 82 | |
| BT026473.1 | - | 4 - 5 | 418 | 82 | ||
More Information
Set Membership: |
|

La2
Genomic Map