Assay Details
| Gene Symbol: La2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001334930.1 | NP_001319421.1 | 6 - 7 | 850 | 99 | |
| NM_106637.5 | NP_178106.2 | 7 - 8 | 775 | 99 | ||
| NM_202456.2 | NP_974185.1 | 6 - 7 | 809 | 99 | ||
| GenBank mRNA | AK319137.1 | - | 6 - 7 | 810 | 99 | |
| BT026473.1 | - | 7 - 8 | 703 | 99 | ||
More Information
Set Membership: |
|

La2
Genomic Map