Assay Details
| Gene Symbol: APT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_106654.3 | NP_178122.1 | 1 - 2 | 841 | 76 | |
| GenBank mRNA | AY072021.1 | - | 1 - 2 | 155 | 76 | |
| BT001122.1 | - | 1 - 2 | 92 | 76 | ||
| X96866.1 | - | 114 | 76 | |||
More Information
Set Membership: |
|

APT2
Genomic Map