Assay Details
| Gene Symbol: APT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001334951.1 | NP_001320554.1 | 3 - 4 | 756 | 65 | |
| NM_106654.3 | NP_178122.1 | 4 - 5 | 1198 | 65 | ||
| GenBank mRNA | AY072021.1 | - | 4 - 5 | 512 | 65 | |
| BT001122.1 | - | 4 - 5 | 449 | 65 | ||
| X96866.1 | - | 471 | 65 | |||
More Information
Set Membership: |
|

APT2
Genomic Map