Assay Details
| Gene Symbol: SUS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_106656.4 | NP_178124.2 | 11 - 12 | 3843 | 94 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SUS2| Gene Symbol: SUS2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_106656.4 | NP_178124.2 | 11 - 12 | 3843 | 94 | |
Set Membership: |
|