Assay Details
| Gene Symbol: CBSX4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001334956.1 | NP_001319425.1 | 3 - 4 | 628 | 93 | |
| NM_106658.3 | NP_178126.2 | 2 - 3 | 592 | 93 | ||
| GenBank mRNA | AK118213.1 | - | 3 - 4 | 707 | 93 | |
More Information
Set Membership: |
|

CBSX4
Genomic Map