Assay Details
| Gene Symbol: CBSX4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001334956.1 | NP_001319425.1 | 4 - 5 | 732 | 85 | |
| NM_106658.3 | NP_178126.2 | 3 - 4 | 696 | 85 | ||
| GenBank mRNA | AK118213.1 | - | 4 - 5 | 811 | 85 | |
More Information
Set Membership: |
|

CBSX4
Genomic Map