Assay Details
| Gene Symbol: MOT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_106680.3 | NP_178147.1 | 1 | 1839 | 98 | |
| GenBank mRNA | AY064056.1 | - | 1 | 1529 | 98 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

MOT2| Gene Symbol: MOT2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_106680.3 | NP_178147.1 | 1 | 1839 | 98 | |
| GenBank mRNA | AY064056.1 | - | 1 | 1529 | 98 | |
Set Membership: |
|