Assay Details
| Gene Symbol: ERH3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_106684.5 | NP_178151.1 | 5 - 6 | 1421 | 126 | |
| GenBank mRNA | AF048706.1 | - | 5 - 6 | 1212 | 126 | |
| AF358779.1 | - | 5 - 6 | 1225 | 126 | ||
| AK228515.1 | - | 5 - 6 | 1358 | 126 | ||
| AY059919.1 | - | 5 - 6 | 1358 | 126 | ||
| BT000149.1 | - | 5 - 6 | 1203 | 126 | ||
More Information
Set Membership: |
|

ERH3
Genomic Map