Assay Details
| Gene Symbol: EMB2753 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001198519.1 | NP_001185448.1 | 17 - 18 | 1853 | 68 | |
| NM_106690.6 | NP_178157.2 | 17 - 18 | 1955 | 68 | ||
| GenBank mRNA | AK118131.1 | - | 17 - 18 | 1848 | 68 | |
More Information
Set Membership: |
|

EMB2753
Genomic Map