Assay Details
| Gene Symbol: IMD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_106704.4 | NP_178171.1 | 4 - 5 | 603 | 99 | |
| GenBank mRNA | Y10216.1 | - | 576 | 99 | ||
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

IMD2| Gene Symbol: IMD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_106704.4 | NP_178171.1 | 4 - 5 | 603 | 99 | |
| GenBank mRNA | Y10216.1 | - | 576 | 99 | ||
Set Membership: |
|