Assay Details
| Gene Symbol: RTL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001334996.1 | NP_001322658.1 | 1 - 2 | 227 | 89 | |
| NM_001334997.1 | NP_001322659.1 | 1 - 2 | 198 | 89 | ||
| NM_001334998.1 | NP_001322660.1 | 1 - 2 | 198 | 89 | ||
| NM_106713.4 | NP_178180.1 | 1 - 2 | 198 | 89 | ||
| GenBank mRNA | AK117343.1 | - | 1 - 2 | 142 | 89 | |
| BT005219.1 | - | 1 - 2 | 111 | 89 | ||
More Information
Set Membership: |
|

RTL1
Genomic Map