Assay Details
Gene Symbol: RTL1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001334996.1 | NP_001322658.1 | 2 - 3 | 424 | 88 | |
NM_001334997.1 | NP_001322659.1 | 2 - 3 | 395 | 88 | ||
NM_001334998.1 | NP_001322660.1 | 2 - 3 | 395 | 88 | ||
NM_106713.4 | NP_178180.1 | 2 - 3 | 395 | 88 | ||
GenBank mRNA | AK117343.1 | - | 2 - 3 | 339 | 88 | |
BT005219.1 | - | 2 - 3 | 308 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
