Assay Details
| Gene Symbol: SAR3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_106716.4 | NP_178183.2 | 4 - 5 | 3181 | 74 | |
| GenBank mRNA | AF411839.1 | - | 4 - 5 | 3055 | 74 | |
| AY942798.1 | - | 4 - 5 | 2844 | 74 | ||
More Information
Set Membership: |
|

SAR3
Genomic Map