Assay Details
| Gene Symbol: TH9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001084655.2 | NP_001078124.1 | 4 - 5 | 559 | 121 | |
| NM_001337765.1 | NP_001325992.1 | 3 - 4 | 738 | 121 | ||
| NM_111705.4 | NP_187483.1 | 4 - 5 | 660 | 121 | ||
| GenBank mRNA | AK226711.1 | - | 4 - 5 | 551 | 121 | |
| BT011728.1 | - | 3 - 4 | 265 | 121 | ||
More Information
Set Membership: |
|

TH9
Genomic Map