Assay Details
| Gene Symbol: LTP6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_111711.4 | NP_187489.1 | 1 - 2 | 429 | 128 | |
| GenBank mRNA | AF159803.1 | - | 1 - 2 | 357 | 128 | |
| AY059951.1 | - | 1 - 2 | 365 | 128 | ||
| AY081617.1 | - | 1 - 2 | 325 | 128 | ||
| AY086646.1 | - | 1 - 2 | 369 | 128 | ||
More Information
Set Membership: |
|

LTP6
Genomic Map